tri-Calciumphosphat, TECHNICAL

Lieferant: VWR Chemicals

Synonyme: Calciumphosphat tribasischCalciumphosphat

22417.293EA 0 EUR
22417.293
tri-Calciumphosphat, TECHNICAL
tri-Calciumphosphat
Formel: Ca₃(PO₄)₂
Molecular Weight: 310,18 g/mol
Dichte: 3,14 g/cm³ (20 °C)
MDL: MFCD00015984
CAS-Nummer: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR