Tetrabutylammoniumperchlorat ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Tetra-n-butylammonium perchlorate

30801.14 30801.22 30801.88
30801.14EA 111 EUR
30801.14 30801.22 30801.88
Tetrabutylammoniumperchlorat ≥99%
Tetrabutylammonium perchlorate

Purity: 99+% CAS No.: 1923-70-2 Molecular Formula: C16H36ClNO4 Formula Weight: 341.92 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: 210-217° UN No.: 1481 MDL No.: MFCD00038722

Formel: C₁₆H₃₆ClNO₄
Molecular Weight: 341,92 g/mol
Schmelzpunkt: 210…217 °C
Dichte: 1,07 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00038722
CAS-Nummer: 1923-70-2
EINECS: 217-655-5
UN: 1481
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR