Tetrabutylammoniumperchlorat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetra-n-butylammonium perchlorate

86885-100G 86885-25G
SIAL86885-100GEA 242 EUR
SIAL86885-100G SIAL86885-25G
Tetrabutylammoniumperchlorat, Sigma-Aldrich®
Tetrabutylammonium perchlorate
Formel: C₁₆H₃₆ClNO₄
Molecular Weight: 341,92 g/mol
Schmelzpunkt: 210 °C
Dichte: 1,07 g/cm³ (20 °C)
MDL: MFCD00038722
CAS-Nummer: 1923-70-2
EINECS: 217-655-5
UN: 1481
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR