Triphenylphosphinsulfid 99+%

Lieferant: Thermo Scientific
Achtung

ACRO159030100EA 44.3 EUR
ACRO159030100
Triphenylphosphinsulfid 99+%
Triphenylphosphine sulphide
Formel: C₁₈H₁₅PS
Molecular Weight: 294,36 g/mol
Schmelzpunkt: 162…164 °C
MDL: MFCD00004917
CAS-Nummer: 3878-45-3
EINECS: 223-407-7
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 162 to 164 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR