Triphenylphosphinsulfid
Lieferant: CHEMPUR
|
Achtung
|
CHMPMB00227.250EA
0
EUR
CHMPMB00227.250
Triphenylphosphinsulfid
Triphenylphosphine sulphide
Formel:
C₁₈H₁₅PS Molecular Weight: 294,36 g/mol Schmelzpunkt: 162…164 °C Dichte: 1,191 g/cm³ (25 °C) |
MDL:
MFCD00004917 CAS-Nummer: 3878-45-3 EINECS: 223-407-7 UN: 3464 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...