Ammoniummetavanadat

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: AmmoniumvanadiumtrioxidAmmoniummonovanadat

HONE14201.9050EA 0 EUR
HONE14201.9050
Ammoniummetavanadat
Ammonium metavanadate
Formel: NH₄VO₃
Molecular Weight: 116,98 g/mol
Schmelzpunkt: 200 °C
Dichte: 2,33 g/cm³ (20 °C)
MDL: MFCD00011430
CAS-Nummer: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1,II
Merck Index: 13,00572

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR