Titancarbid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL594849-25GEA
71.8
EUR
SIAL594849-25G
Titancarbid, Sigma-Aldrich®
Titanium carbide
Formel:
CTi Molecular Weight: 59,89 g/mol Siedepunkt: 4300 °C (1013 hPa) Schmelzpunkt: 3067 °C Dichte: 4,93 g/cm³ (25 °C) |
MDL:
MFCD00011268 CAS-Nummer: 12070-08-5 EINECS: 235-120-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...