Hafnium(IV)chlorid ≥99,9% (Metall-Basis) (max. 0,5% Zr)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Hafnium tetrachloride
45612.14 45612.22 45612.36
45612.14EA 90.9 EUR
45612.14 45612.22 45612.36
Hafnium(IV)chlorid ≥99,9% (Metall-Basis) (max. 0,5% Zr)
Hafnium(IV)chlorid
Purity: 99.9% CAS No.: 13499-05-3 Molecular Formula: Cl4Hf Formula Weight: 320.30 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Odour: Irritating Melting Point: 320° subl. Merck Reference: 14,4588 UN No.: 3260 MDL No.: MFCD00011034
Formel: Cl₄Hf
Molecular Weight: 320,3 g/mol
Schmelzpunkt: 320 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011034
CAS-Nummer: 13499-05-3
EINECS: 236-826-5
UN: 3260
ADR: 8,II
Merck Index: 14,4588