Pyronin G

Lieferant: POLYSCIENCES EUROPE

Synonyme: Pyronin Y3,6-Bis(dimethylamino)xanthyliumchlorid

POLS18614-5EA 241 EUR
POLS18614-5
Pyronin G
Pyronin G
Formel: C₁₇H₁₉ClN₂O
Molecular Weight: 302,8 g/mol
Schmelzpunkt: 250…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011725
CAS-Nummer: 92-32-0
EINECS: 202-147-8
Merck Index: 12,08190

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR