Centrifuge for oil tests, Seta Oil
Lieferant: Stanhope Seta
Seta Oil
752-0000EA 0 EUR
752-0000
Centrifuge for oil tests, Seta Oil
Zentrifugen Tischzentrifugen
Seta Oil Test centrifuge is a microprocessor controlled, heated centrifuge. The instrument is fully programmable allowing automatic configuration to the specified oil test parameters.
  • Static, almost vertical bucket positioning
  • Membrane ‘touch’ control panel
  • Pre-heat facility to ensure bowl is at test temperature prior to test
  • Universal buckets for 12,5 ml, 6 and 8 inch glassware
  • Microprocessor control with memory for 99 run profiles
  • Optional ‘Setafuge’ hazard reduction system
Standards: Conforms to safety standards BS 4402, DIN 58970, IEC 1010-1, IEC 1010-2 D; pr EN 61 010-1 and 61 010-1; BGR 500, Chapter 2.11, Part 3; UVV VBG 4, and VDE 0871.
Bestellinformation: Supplied with 4-place swing-out rotor, buckets and cushions. Tubes, adapters and the imbalance micro switch need to be ordered separately.