Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Lieferant: Strem Chemicals, Inc.
STRE44-6040EA 299 EUR
STRE44-6040
Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Formel: C₃₀H₃₆Cl₂N₂P₂Ru
Molecular Weight: 658,54 g/mol
MDL: MFCD15144861
CAS-Nummer: 911128-25-1