Safranin O acc. Gram-Hücker

Lieferant: RAL (REACTIFS RAL SA)

Synonyme: Basisrot 23,7-Diamino-2,8-dimethyl-5-phenylphenaziniumchlorid

720-1163EA 50.44 EUR
720-1163
Safranin O acc. Gram-Hücker
Safranin O

Used in Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria.


Used in Gram-Hücker R Kit Ref 361520


Gram-Hücker staining is based on the same principle than the Gram-Nicolle staining. The structure of the bacterial walls and their permeability to alcohol help differentiate the Gram-negative bacteria from the Gram-positive ones. In this technique, Carbolic Gentian Violet is replaced by Crystal Violet Oxalate (that does not require filtration) and Ziehl-Carbolic Fuchsin by Safranin.

Formel: C₂₀H₁₉ClN₄
Molecular Weight: 350,85 g/mol
MDL: MFCD00011759
CAS-Nummer: 477-73-6
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR