Lacidipine

Lieferant: Thermo Scientific
Achtung

ACRO463180010EA 246 EUR
ACRO463180010
Lacidipine
Lacidipine

  • Bioactive

Formel: C₂₆H₃₃NO₆
Molecular Weight: 455,55 g/mol
Schmelzpunkt: 179 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00865936
CAS-Nummer: 103890-78-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 177 to 181 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR