Naphthol AS-BI-phosphat-Lösung 4 g/L in Methanol stabilisiert, Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
Gefahr
M-Clarity™ Qualitätsstufe = MQ500
1802-50MLEA 146 EUR
1802-50ML
Naphthol AS-BI-phosphat-Lösung 4 g/L in Methanol stabilisiert, Sigma-Aldrich®
Naphthol AS-BI-phosphat
Naphthol AS-BI Phosphoric acid solution for use in the Sigma Lymphocyte Enzyme (Acid Phosphatase) Procedure, No. 181A.
  • Suitable for haematology
  • Suitable for histology
Formel: C₁₈H₁₅BrNO₆P
Molecular Weight: 452,1926 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00004058
CAS-Nummer: 1919-91-1
UN: 1993
ADR: 3,III
IVDLogo