Fast Dark Blue R Salt, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY

F0750-25GEA 54.4 EUR
F0750-25G
Fast Dark Blue R Salt, Sigma-Aldrich®
Fast Dark Blue R Salt

Fast Dark Blue R Salt, (Azoic Diazo Component 51; Azoic Diazo No. 51)

Formel: C₁₄HCl₂N₅O₄
Molecular Weight: 486,77 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00135879
CAS-Nummer: 6709-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR