Hämatoxylin Gill No. 3, Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
Gefahr
Synonyme: HydroxybrasilinNaturschwarz 1
GHS316-500MLEA 74.6 EUR
GHS316-500ML
Hämatoxylin Gill No. 3, Sigma-Aldrich®
Hämatoxylin-Färbelösung
Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time. Used with hematoxylin and eosin staining.
Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078111
CAS-Nummer: 517-28-2
IVDLogo