Fluorescein-5-EX N-hydroxysuccinimide ester, Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
F9551-10MGEA
429
EUR
F9551-10MG
Fluorescein-5-EX N-hydroxysuccinimide ester, Sigma-Aldrich®
Fluorescein-5-EX N-hydroxysuccinimide ester
Fluorescein-5-EX N-hydroxysuccinimide ester has been used for the labeling of antibodies.