8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY
Achtung

Synonyme: APTS

A7222-10MGEA 430 EUR
A7222-10MG
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt, Sigma-Aldrich®
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt

Fluorescent label for glycoproteins and saccharides.
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt (APTS) has been used for the detection of human milk oligosaccharides using multiplexed CGE with LIF (capillary gel electrophoresis with laser-induced

Formel: C₁₆H₈NNa₃O₉S₃
Molecular Weight: 523,40 g/mol
Schmelzpunkt: ≥250 °C (Lit.)
Lagertemperatur: Kühlschrank
MDL: MFCD00674494
CAS-Nummer: 196504-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR