Quin 2 potassium salt hydrate, Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
Q0501-10MGEA
536
EUR
Q0501-10MG
Quin 2 potassium salt hydrate, Sigma-Aldrich®
Quin 2 potassium salt hydrate
Calcium-specific chelator and indicator. Upon binding calcium there is a shift in the UV spectrum and an enhancement of fluorescence.
Formel:
C₂₆H₂₇N₃O Molecular Weight: 559,52 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00167201 CAS-Nummer: 149022-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...