Quinoline yellow 95% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
|
Achtung
|
234133-25GEA
68.2
EUR
234133-25G
Quinoline yellow 95% (Farbstoffgehalt), Sigma-Aldrich®
Quinoline yellow
Quinoline Yellow, Dye content 95 % (Solvent yellow 33)
Formel:
C₁₈H₁₁NO₂ Molecular Weight: 273,29 g/mol Schmelzpunkt: 240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011593 CAS-Nummer: 8003-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...