Brilliant black BN, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY

211842-10GEA 93 EUR
211842-10G
Brilliant black BN, Sigma-Aldrich®
Brilliant black BN

Brilliant Black BN has been used: in micronucleus and comet assays to evaluate its genotoxic effect.as a quenching agent in calcium mobilization assay.

Formel: C₂₈H₁₇N₅O₁₄S₄
Molecular Weight: 867,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078980
CAS-Nummer: 2519-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR