Azur II eosinate, Pulver, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Azure II eosin, Dipotassium 7-(dimethylamino)-N,N-dimethylphenothiazin-3-iminium N,N-dimethyl-7-(methylamino)phenothiazin-3-iminium eosin YS(2-) dichloride, Azure II eosinate
M-Clarity™ Qualitätsstufe = MQ100
200220-100G
200220-25G
200220-100GEA
396
EUR
200220-100G
200220-25G
Azur II eosinate, Pulver, Sigma-Aldrich®
Azur II eosinate
Chief ingredient of Giemsa Stain.
Formel:
C₅₁H₄₀Br₄Cl₂N₆Na₂O₅S₂ Molecular Weight: 1317,534 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00084753 CAS-Nummer: 53092-85-6 |
Spezifikation Testergebnisse
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Wavelength C=0.005 G/L Methanol | 645 - 655 nm |
Extinction Coefficient | ≥49440 |
Wavelength | 243 - 247 nm |
Solubility (Turbidity) | Hazy to Turbid |
Solubility (Color) | Dark Blue |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...