4-Aminobenzaldehyde, N-BOC protected 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(4-Formyl-phenyl)-carbamicacidtert-butylester, tert-Butyl (4-formylphenyl)carbamate
OR01705-5G
APOSOR01705-5GEA
156
EUR
APOSOR01705-5G
4-Aminobenzaldehyde, N-BOC protected 97%
4-Aminobenzaldehyde, N-BOC protected
Formel:
C₁₂H₁₅NO₃ Molecular Weight: 221,252 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245541 CAS-Nummer: 144072-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...