4-(2-Aminoethyl)piperidine, 4-BOC protected

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(2-Boc-aminoethyl)piperidine, tert-Butyl [2-(piperidin-4-yl)ethyl]carbamate, 4-[2-(Boc-amino)ethyl]piperidine, N-(tert-Butoxycarbonyl)-2-(piperidin-4-yl)ethylamine

OR2257-1G
APOSOR2257-1GEA 56.7 EUR
APOSOR2257-1G
4-(2-Aminoethyl)piperidine, 4-BOC protected
4-(2-Aminoethyl)piperidine, 4-BOC protected
Formel: C₁₂H₂₄N₂O₂
Molecular Weight: 228,331 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318373
CAS-Nummer: 165528-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR