Allylamine, N-CBZ protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzyl (prop-2-en-1-yl)carbamate, Benzyl allylcarbamate
OR28285-1G
OR28285-250MG
APOSOR28285-1GEA
244
EUR
APOSOR28285-1G
APOSOR28285-250MG
Allylamine, N-CBZ protected
Allylamine, N-CBZ protected
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191,226 g/mol Siedepunkt: 120…125 °C (1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570995 CAS-Nummer: 5041-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...