3-((E)-2-Dimethylaminovinyl)isonicotinonitrile 95+%
Lieferant: Apollo Scientific
APOSOR346511-1GEA
412
EUR
APOSOR346511-1G
3-((E)-2-Dimethylaminovinyl)isonicotinonitrile 95+%
3-((E)-2-Dimethylaminovinyl)isonicotinonitrile
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,214 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13181594 CAS-Nummer: 601514-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...