Azocarmine G

Lieferant: Apollo Scientific
Achtung

Synonyme: Rosinduline, Acid red 101, Basalan Carmine GE

BIA1501-10G BIA1501-25G
APOSBIA1501-10GEA 145 EUR
APOSBIA1501-10G APOSBIA1501-25G
Azocarmine G
Azocarmine G
Formel: C₂₈H₁₇N₃Na₂O₆S₂
Molecular Weight: 601,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042002
CAS-Nummer: 25641-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR