Methylenblau, BioChemica (C.I. 52015)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methylthioniniumchlorid
, 3,7-Bis(dimethylamino)phenothiaziniumchlorid
, N,N,N′,N′-Tetramethylthioninchlorid
APOSBIA4084-100GEA
198
EUR
APOSBIA4084-100G
Methylenblau, BioChemica (C.I. 52015)
Methylenblau
Formel:
C₁₆H₁₈ClN₃S Molecular Weight: 319,86 g/mol Schmelzpunkt: 180 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150008 CAS-Nummer: 61-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...