Phomopsin A

Lieferant: Apollo Scientific

BIA4325-1MG BIA4325-5MG
APOSBIA4325-1MGEA 487 EUR
APOSBIA4325-1MG APOSBIA4325-5MG
Phomopsin A
Phomopsin A

Phomopsin A mycotoxin extracted from Phomopsis leptostromiformis.


  • Clear colourless solution at 10mg/ml in DMSO & 5 mg/ml in Methanol.

Formel: C₃₆H₄₅ClN₆O₁₂
Molecular Weight: 789,2 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 64925-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR