6-Iodo diosmin 98% min

Lieferant: Apollo Scientific

BICL4116-1MG BICL4116-2MG
APOSBICL4116-1MGEA 434 EUR
APOSBICL4116-1MG APOSBICL4116-2MG
6-Iodo diosmin 98% min
6-Iodo diosmin
Formel: C₂₈H₃₁IO₁₅
Molecular Weight: 734,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28515355
CAS-Nummer: 1431536-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR