D(-)-Arabinose 99%

Lieferant: Apollo Scientific
Achtung

APOSBIFS0101-1KGEA 285 EUR
APOSBIFS0101-1KG APOSBIFS0101-500G APOSBIFS0101-100G
D(-)-Arabinose 99%
D(-)-Arabinose
Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Schmelzpunkt: 153…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135608
CAS-Nummer: 10323-20-3
EINECS: 233-708-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR