Hygromycin B, Pulver

Lieferant: Apollo Scientific
Gefahr

APOSBIH4302-1GEA 241 EUR
APOSBIH4302-1G
Hygromycin B, Pulver
Hygromycin B

Toxic aminoglycoside produced by Streptomyces hygroscopicus. Used as a selective agent for the incorporation of the APH-4 gene in plant tissue.


  • Average Activity: 1000 units/mg
  • Cell culture tested

Formel: C₂₀H₃₇N₃O₁₃
Molecular Weight: 527,5 g/mol
Schmelzpunkt: 160…180 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD06795479
CAS-Nummer: 31282-04-9
UN: 3462
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR