Phosphoenolpyruvic acid monocyclohexylammonium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: PEP·CHX

APOSBIP0688-5GEA 102 EUR
APOSBIP0688-5G APOSBIP0688-1G
Phosphoenolpyruvic acid monocyclohexylammonium salt
Phosphoenolpyruvic acid monocyclohexylammonium salt
Formel: C₃H₅O₆P
Molecular Weight: 267,22 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036375
CAS-Nummer: 10526-80-4
EINECS: 234-084-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR