Puromycin Dihydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

BIP3340-100MG BIP3340-250MG BIP3340-500MG
APOSBIP3340-100MGEA 132 EUR
APOSBIP3340-100MG APOSBIP3340-250MG APOSBIP3340-500MG
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid
Formel: C₂₂H₂₉N₇O₅·2HCl
Molecular Weight: 544,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012691
CAS-Nummer: 58-58-2
EINECS: 200-387-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR