Staurosporine (aus Streptomyces species)

Lieferant: Apollo Scientific
Gefahr

Synonyme: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282

BIS0504-10MG BIS0504-25MG BIS0504-100MG
APOSBIS0504-10MGEA 87.3 EUR
APOSBIS0504-10MG APOSBIS0504-25MG APOSBIS0504-100MG
Staurosporine (aus Streptomyces species)
Staurosporine

Potent inhibitor of phospholipid/calcium-dependent.protein kinase.

Formel: C₂₈H₂₆N₄O₃
Molecular Weight: 466,541 g/mol
Schmelzpunkt: 270 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD18252446
CAS-Nummer: 62996-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR