Xanthydrol

Lieferant: Apollo Scientific

Synonyme: 9-Hydroxyxanthen9H-Xanthen-9-ol9-Xanthenol

APOSBIX1001-5GEA 68.4 EUR
APOSBIX1001-5G APOSBIX1001-25G
Xanthydrol
Xanthydrol
Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
Schmelzpunkt: 124…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005057
CAS-Nummer: 90-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR