Hafnium(IV)chlorid 98+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Hafnium tetrachloride
IN1954-100G
IN1954-25G
APOSIN1954-100GEA
0
EUR
APOSIN1954-100G
APOSIN1954-25G
Hafnium(IV)chlorid 98+%
Hafnium(IV)chlorid
Formel:
Cl₄Hf Molecular Weight: 320,3 g/mol Schmelzpunkt: 320 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011034 CAS-Nummer: 13499-05-3 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...