Lanthan(III)chlorid Heptahydrat ACS
Lieferant: Apollo Scientific
|
Achtung
|
APOSIN2120-100GEA
121
EUR
APOSIN2120-100G
Lanthan(III)chlorid Heptahydrat ACS
Lanthan(III)chlorid Heptahydrat
Formel:
LaCl₃·7H₂O Molecular Weight: 371,37 g/mol Schmelzpunkt: 91 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149756 CAS-Nummer: 10025-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...