1,3-Propandiol

Lieferant: Apollo Scientific

Synonyme: Trimethylenglycol1,3-Propylenglycol1,3-Dihydroxypropan

APOSOR0974-250GEA 109 EUR
APOSOR0974-250G APOSOR0974-1KG
1,3-Propandiol
1,3-Propandiol
Formel: HO(CH₂)₃OH
Molecular Weight: 76,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002949
CAS-Nummer: 504-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR