(1-Ethoxycarbonylethyliden)triphenylphosphoran 95%

Lieferant: Apollo Scientific
Achtung

APOSOR10643-500GEA 0 EUR
APOSOR10643-500G APOSOR10643-1KG APOSOR10643-100G
(1-Ethoxycarbonylethyliden)triphenylphosphoran 95%
(1-Ethoxycarbonylethyliden)triphenylphosphoran
Formel: C₂₃H₂₃O₂P
Molecular Weight: 362,41 g/mol
Schmelzpunkt: 157…159 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009160
CAS-Nummer: 5717-37-3
EINECS: 227-215-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR