4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

APOSOR110196-1GEA 126 EUR
APOSOR110196-1G
4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde
4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde
Formel: C₅H₅BrN₂O
Molecular Weight: 189,02 g/mol
Schmelzpunkt: 97…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03074341
CAS-Nummer: 473528-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR