4-Nitrobenzaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR12008-500GEA
150
EUR
APOSOR12008-500G
APOSOR12008-100G
4-Nitrobenzaldehyd 95%
4-Nitrobenzaldehyd
Formel:
C₇H₅NO₃ Molecular Weight: 151,12 g/mol Schmelzpunkt: 103…106 °C Dichte: 1,496 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007346 CAS-Nummer: 555-16-8 EINECS: 209-084-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...