4'-Chlorchalkon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Chlorbenzylidenacetophenon
APOSOR1381-5GEA
14.7
EUR
APOSOR1381-5G
APOSOR1381-25G
4'-Chlorchalkon 98%
4'-Chlorchalkon
Formel:
C₁₅H₁₁ClO Molecular Weight: 242,7 g/mol Schmelzpunkt: 97…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016343 CAS-Nummer: 956-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...