2-Amino-3-brom-5-methylpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-5-methyl-2-pyridylamin2-Amino-3-brom-5-picolin3-Brom-5-methylpyridin-2-amin

APOSOR1651-10GEA 14.7 EUR
APOSOR1651-10G
2-Amino-3-brom-5-methylpyridin
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR