3-Amino-2-brom-6-methylpyridin

Lieferant: Apollo Scientific
Achtung

APOSOR2557-5GEA 120 EUR
APOSOR2557-5G APOSOR2557-10G APOSOR2557-1G
3-Amino-2-brom-6-methylpyridin
3-Amino-2-bromo-6-methylpyridine
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03095078
CAS-Nummer: 126325-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR