Pivalamidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR27177-25GEA
265
EUR
APOSOR27177-25G
APOSOR27177-1G
APOSOR27177-10G
Pivalamidine hydrochloride
Pivalamidine hydrochloride
Formel:
C₅H₁₂N₂ Molecular Weight: 136,62 g/mol Schmelzpunkt: 184…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051988 CAS-Nummer: 18202-73-8 EINECS: 242-091-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...