3-Amino-5-methylisoxazol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methylisoxazol-3-ylamin5-Methylisoxazol-3-amin

APOSOR28614-25GEA 102 EUR
APOSOR28614-25G
3-Amino-5-methylisoxazol 97%
3-Amino-5-methylisoxazole
Formel: C₄H₆N₂O
Molecular Weight: 98,1 g/mol
Schmelzpunkt: 61…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003155
CAS-Nummer: 1072-67-9
EINECS: 214-013-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR