1-Amino-7-bromisochinolin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromoisoquinolin-1-amine

OR300055-1G OR300055-250MG OR300055-5G
APOSOR300055-1GEA 346 EUR
APOSOR300055-1G APOSOR300055-250MG APOSOR300055-5G
1-Amino-7-bromisochinolin 97%
1-Amino-7-bromoisoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374397
CAS-Nummer: 215453-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR