8-Bromoisoquinolin-1(2H)-one

Lieferant: Apollo Scientific
Achtung

OR300126-1G OR300126-250MG
APOSOR300126-1GEA 120 EUR
APOSOR300126-1G APOSOR300126-250MG
8-Bromoisoquinolin-1(2H)-one
8-Bromoisoquinolin-1(2H)-one
Formel: C₉H₆BrNO
Molecular Weight: 224,0540 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858429
CAS-Nummer: 475994-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR