2-Chlor-3-methylpyrazin

Lieferant: Apollo Scientific

APOSOR301125-1GEA 61.1 EUR
APOSOR301125-1G
2-Chlor-3-methylpyrazin
2-Chloro-3-methylpyrazine
Formel: C₅H₅ClN₂
Molecular Weight: 128,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023279
CAS-Nummer: 95-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR